(R)-2-(4'-(4-(((1-(2-Chlorophenyl)ethoxy)carbonyl)amino)-3-methylisoxazol-5-yl)-[1,1'-biphenyl]-4-yl)acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: AM966

A234317-10MG A234317-1MG A234317-2MG A234317-5MG A234317-250MG A234317-100MG A234317-1G A234317-50MG
77165-642EA 57.68 USD
77165-642 77165-644 77165-646 77165-648 77612-292 77683-686 77683-687 77683-688
(R)-2-(4'-(4-(((1-(2-Chlorophenyl)ethoxy)carbonyl)amino)-3-methylisoxazol-5-yl)-[1,1'-biphenyl]-4-yl)acetic acid 98%
(R)-2-(4'-(4-(((1-(2-Chlorophenyl)ethoxy)carbonyl)amino)-3-methylisoxazol-5-yl)-[1,1'-biphenyl]-4-yl)acetic acid

Precaución: For research use only.

Formula: C₂₇H₂₃ClN₂O₅
MW: 490.93 g/mol
MDL Number: MFCD20486575
CAS Number: 1228690-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR