(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2,4-difluorophenyl)propanoic acid 98%

Supplier: Ambeed
Warning

A234238-100MG A234238-1G A234238-250MG
77165-636EA 27.81 USD
77165-636 77165-638 77165-640
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2,4-difluorophenyl)propanoic acid 98%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2,4-difluorophenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₂₄H₁₉F₂NO₄
MW: 423.41 g/mol
MDL Number: MFCD07371998
CAS Number: 1032337-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR