Methyl (4-bromo-3-fluorophenyl)acetate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-(4-bromo-3-fluorophenyl)acetate

A234233-5G A234233-1G A234233-250MG
77165-634EA 126 USD
77165-634 77892-397 77892-398
Methyl (4-bromo-3-fluorophenyl)acetate 98%
Methyl (4-bromo-3-fluorophenyl)acetate

Precaución: for Research Use Only

Formula: C₉H₈BrFO₂
MDL Number: MFCD22418249
CAS Number: 942282-41-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR