Benzyl 2-amino-2-methylpropanoate hydrochloride 98%

Supplier: Ambeed
Warning

A234187-5G A234187-25G
77193-166EA 23 USD
77193-166 77568-274
Benzyl 2-amino-2-methylpropanoate hydrochloride 98%
Benzyl 2-amino-2-methylpropanoate hydrochloride

Precaución: For research use only.

Formula: C₁₁H₁₆ClNO₂
MDL Number: MFCD14155677
CAS Number: 60421-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR