Ethyl 3-(2,6-Difluorophenyl)-3-oxopropionate 97%

Supplier: Ambeed
Warning

A233956-100MG A233956-250MG
77165-622EA 6 USD
77165-622 77165-624
Ethyl 3-(2,6-Difluorophenyl)-3-oxopropionate 97%
Ethyl 3-(2,6-Difluorophenyl)-3-oxopropionate

Precaución: for Research Use Only

Formula: C₁₁H₁₀F₂O₃
MDL Number: MFCD11041490
CAS Number: 97305-12-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR