2-(3-Fluorophenyl)ethanol 98%

Supplier: Ambeed
Warning

A233825-1G A233825-25G A233825-5G
77165-618EA 5.15 USD
77165-618 77165-620 77683-685
2-(3-Fluorophenyl)ethanol 98%
2-(3-Fluorophenyl)ethanol

Precaución: For research use only.

Formula: C₈H₉FO
MW: 140.16 g/mol
MDL Number: MFCD00045998
CAS Number: 52059-53-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR