6-Methylimidazo[1,2-a]pyridine-2-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
A233418-100G
A233418-10G
A233418-1G
A233418-250MG
A233418-25G
A233418-5G
77165-604EA
1608.86
USD
77165-604
77165-606
77165-608
77165-610
77165-612
77165-614
6-Methylimidazo[1,2-a]pyridine-2-carboxylic acid 97%
6-Methylimidazo[1,2-a]pyridine-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₉H₈N₂O₂ MW: 176.1700 g/mol |
MDL Number:
MFCD03425068 CAS Number: 80353-93-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...