Methyl 3-(hydroxymethyl)bicyclo[1.1.1]pentane-1-carboxylate 98%
Supplier: Ambeed
|
Warning
|
A232942-100MG
A232942-1G
A232942-5G
A232942-250MG
77165-586EA
113.3
USD
77165-586
77568-258
77568-260
77612-278
Methyl 3-(hydroxymethyl)bicyclo[1.1.1]pentane-1-carboxylate 98%
Methyl 3-(hydroxymethyl)bicyclo[1.1.1]pentane-1-carboxylate
Precaución: For research use only.
Formula:
C₈H₁₂O₃ MW: 156.179 g/mol |
MDL Number:
MFCD23164154 CAS Number: 180464-87-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...