1-(6-Bromopyridin-2-yl)-2-methylpropan-2-ol 95%

Supplier: Ambeed
Warning

A232399-1G A232399-250MG
77193-152EA 117 USD
77193-152 77193-154
1-(6-Bromopyridin-2-yl)-2-methylpropan-2-ol 95%
1-(6-Bromopyridin-2-yl)-2-methylpropan-2-ol

Precaución: for Research Use Only

Formula: C₉H₁₂BrNO
MDL Number: MFCD11977453
CAS Number: 955369-59-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR