BLU9931 99%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: N-(2-((6-(2,6-Dichloro-3,5-dimethoxyphenyl)quinazolin-2-yl)amino)-3-methylphenyl)acrylamide

A231990-10MG A231990-1MG A231990-5MG A231990-100MG A231990-50MG A231990-250MG
77165-544EA 69 USD
77165-544 77165-546 77165-548 77612-266 77612-268 77683-675
BLU9931 99%, Ambeed.Inc
BLU9931

Precaución: For research use only.

Formula: C₂₆H₂₂Cl₂N₄O₃
MW: 509.38 g/mol
MDL Number: MFCD28900729
CAS Number: 1538604-68-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR