INCB39110 98+%

Supplier: Ambeed
Warning

Synonyms: Itacitinib, 2-(3-(4-(7H-pyrrolo[2,3-d]pyrimidin-4-yl)-1H-pyrazol-1-yl)-1-(1-(3-fluoro-2-(trifluoromethyl)isonicotinoyl)piperidin-4-yl)azetidin-3-yl)acetonitrile, INCB039110

A231782-10MG A231782-1MG A231782-50MG A231782-5MG
77193-144EA 143 USD
77193-144 77193-146 77193-148 77193-150
INCB39110 98+%
INCB39110

Precaución: for Research Use Only

Formula: C₂₆H₂₃F₄N₉O
MW: 553.51 g/mol
MDL Number: MFCD28579595
CAS Number: 1334298-90-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR