tert-Butyl-4-{[(methylsulfonyl)oxy]methyl}tetrahydro-1(2H)-pyridinecarboxylate 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-(((methylsulphonyl)oxy)methyl)piperidine-1-carboxylate, tert-Butyl 4-([(methylsulphonyl)oxy]methyl)tetrahydro-1(2h)-pyridinecarboxylate

A231118-10G A231118-1G A231118-250MG A231118-5G
77193-136EA 113.3 USD
77193-136 77193-138 77193-140 77193-142
tert-Butyl-4-{[(methylsulfonyl)oxy]methyl}tetrahydro-1(2H)-pyridinecarboxylate 95%
tert-Butyl-4-{[(methylsulfonyl)oxy]methyl}tetrahydro-1(2H)-pyridinecarboxylate

Precaución: for Research Use Only

Formula: C₁₂H₂₃NO₅S
MW: 293.38 g/mol
MDL Number: MFCD02082459
CAS Number: 161975-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR