6-Bromo-2,3-difluoroaniline 95%

Supplier: Ambeed
Warning

A230889-100MG A230889-1G A230889-250MG A230889-5G A230889-25G
77165-522EA 7.21 USD
77165-522 77165-524 77165-526 77165-528 77568-232
6-Bromo-2,3-difluoroaniline 95%
6-Bromo-2,3-difluoroaniline

Precaución: For research use only.

Formula: C₆H₄BrF₂N
MW: 208.003 g/mol
MDL Number: MFCD03840796
CAS Number: 887579-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR