AMG-47a 98%

Supplier: Ambeed
Warning

Synonyms: 4-Methyl-3-(2-((2-morpholinoethyl)amino)quinazolin-6-yl)-N-(3-(trifluoromethyl)phenyl)benzamide

A230809-10MG A230809-1MG A230809-2MG A230809-5MG
77193-128EA 125.66 USD
77193-128 77193-130 77193-132 77193-134
AMG-47a 98%
AMG-47a

Precaución: for Research Use Only

Formula: C₂₉H₂₈F₃N₅O₂
MW: 535.6 g/mol
MDL Number: MFCD09970318
CAS Number: 882663-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR