2-Chloro-1-ethyl-1H-benzo[d]imidazole 97%

Supplier: Ambeed
Warning

A230689-100MG A230689-10G A230689-1G A230689-250MG A230689-5G
77165-512EA 10.3 USD
77165-512 77165-514 77165-516 77165-518 77165-520
2-Chloro-1-ethyl-1H-benzo[d]imidazole 97%
2-Chloro-1-ethyl-1H-benzo[d]imidazole

Precaución: for Research Use Only

Formula: C₉H₉ClN₂
MW: 180.63 g/mol
MDL Number: MFCD00769822
CAS Number: 58533-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR