tert-Butyl 3-ethyl-4-oxopiperidine-1-carboxylate 95%

Supplier: Ambeed
Warning

A230096-100MG A230096-1G A230096-250MG A230096-5G
77193-104EA 25.75 USD
77193-104 77193-106 77193-108 77193-110
tert-Butyl 3-ethyl-4-oxopiperidine-1-carboxylate 95%
tert-Butyl 3-ethyl-4-oxopiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₃
MW: 227.30 g/mol
MDL Number: MFCD08460961
CAS Number: 117565-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR