4'-Fluorobutyrophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Fluorophenyl)butan-1-one

A230089-100G A230089-10G A230089-5G
77193-098EA 92.7 USD
77193-098 77193-100 77193-102
4'-Fluorobutyrophenone 98%
4'-Fluorobutyrophenone

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO
MW: 166.2 g/mol
MDL Number: MFCD00017967
CAS Number: 582-83-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR