4-Bromo-1-methyl-1H-1,3-benzodiazole 95%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-1-methyl-1H-benzo[d]imidazole

A229988-10G A229988-250MG A229988-5G A229988-1G
77165-494EA 358.44 USD
77165-494 77165-496 77165-498 77568-226
4-Bromo-1-methyl-1H-1,3-benzodiazole 95%
4-Bromo-1-methyl-1H-1,3-benzodiazole

Precaución: For research use only.

Formula: C₈H₇BrN₂
MW: 211.07 g/mol
MDL Number: MFCD18378523
CAS Number: 1353679-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR