1-(4-(Aminomethyl)phenyl)-N,N-dimethylmethanamine hydrochloride 95%

Supplier: Ambeed
Warning

A229911-1G A229911-250MG
77165-482EA 87.55 USD
77165-482 77165-484
1-(4-(Aminomethyl)phenyl)-N,N-dimethylmethanamine hydrochloride 95%
1-(4-(Aminomethyl)phenyl)-N,N-dimethylmethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₇ClN₂
MW: 200.70 g/mol
MDL Number: MFCD28134445
CAS Number: 242452-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR