N-(2-Hydroxyethyl)maleimide 97%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Hydroxyethyl)-1H-pyrrole-2,5-dione, 1-(2-Hydroxyethyl)maleimide, N-(Ethanol)maleimide, 1-(2-Hydroxyethyl)-pyrrole-2,5-dione, HEMI

A229302-100MG A229302-1G A229302-250MG A229302-5G A229302-25G
77193-088EA 6 USD
77193-088 77193-090 77193-092 77193-094 77568-220
N-(2-Hydroxyethyl)maleimide 97%
N-(2-Hydroxyethyl)maleimide

Precaución: For research use only.

Formula: C₆H₇NO₃
MW: 141.13 g/mol
MDL Number: MFCD00465266
CAS Number: 1585-90-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR