1-(4-Chlorophenyl)-2-pyrrolidinone 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-(4-Chlorophenyl)pyrrolidin-2-one
A229155-100MG
A229155-1G
A229155-250MG
77193-080EA
39.14
USD
77193-080
77193-082
77193-084
1-(4-Chlorophenyl)-2-pyrrolidinone 95%
1-(4-Chlorophenyl)-2-pyrrolidinone
Precaución: for Research Use Only
Formula:
C₁₀H₁₀ClNO MW: 195.6455 g/mol |
MDL Number:
MFCD00138486 CAS Number: 7661-33-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...