tert-Butyl 5-fluoro-1H-indole-1-carboxylate 98%

Supplier: Ambeed
Warning

A228529-1G A228529-250MG A228529-5G
77193-066EA 115 USD
77193-066 77193-068 77193-070
tert-Butyl 5-fluoro-1H-indole-1-carboxylate 98%
tert-Butyl 5-fluoro-1H-indole-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₄FNO₂
MDL Number: MFCD09870066
CAS Number: 129822-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR