tert-Butyl 2-(2,5-difluorobenzoyl)hydrazinecarboxylate 97%

Supplier: Ambeed
Warning

A228417-1G A228417-250MG A228417-25G A228417-5G
77193-054EA 16.48 USD
77193-054 77193-056 77193-058 77193-060
tert-Butyl 2-(2,5-difluorobenzoyl)hydrazinecarboxylate 97%
tert-Butyl 2-(2,5-difluorobenzoyl)hydrazinecarboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₄F₂N₂O₃
MW: 272.24 g/mol
MDL Number: MFCD23700206
CAS Number: 1079843-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR