4-Methyl-3-nitrobenzenesulfonamide 98%

Supplier: Ambeed
Warning

A228267-100MG A228267-1G A228267-250MG A228267-5G
77193-040EA 16.48 USD
77193-040 77193-042 77193-044 77193-046
4-Methyl-3-nitrobenzenesulfonamide 98%
4-Methyl-3-nitrobenzenesulfonamide

Precaución: for Research Use Only

Formula: C₇H₈N₂O₄S
MW: 216.22 g/mol
MDL Number: MFCD00460611
CAS Number: 6949-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR