6-Chloro-4-iodo-3-hydroxypyridine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Chloro-4-iodo-5-hydroxypyridine, 6-Chloro-4-iodopyridin-3-ol
A227897-100MG
A227897-10G
A227897-250MG
A227897-25G
A227897-5G
A227897-1G
77165-452EA
19.57
USD
77165-452
77165-454
77165-456
77165-458
77165-460
77683-662
6-Chloro-4-iodo-3-hydroxypyridine 97%
6-Chloro-4-iodo-3-hydroxypyridine
Precaución: For research use only.
Formula:
C₅H₃ClINO MW: 255.441 g/mol |
MDL Number:
MFCD11977445 CAS Number: 877133-58-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...