2-(3-Bromophenyl)cyclopropanecarboxylic acid 95%

Supplier: Ambeed
Warning

A227872-100MG A227872-250MG
77165-448EA 41 USD
77165-448 77165-450
2-(3-Bromophenyl)cyclopropanecarboxylic acid 95%
2-(3-Bromophenyl)cyclopropanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₉BrO₂
MDL Number: MFCD11895900
CAS Number: 91445-84-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR