3-Methoxy-1,2-benzisothiazole-1,1-dioxide 97%

Supplier: Ambeed
Warning

A227499-100MG A227499-1G
77193-032EA 63 USD
77193-032 77193-034
3-Methoxy-1,2-benzisothiazole-1,1-dioxide 97%
3-Methoxy-1,2-benzisothiazole-1,1-dioxide

Precaución: for Research Use Only

Formula: C₈H₇NO₃S
MDL Number: MFCD00486244
CAS Number: 18712-14-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR