2-(3,4-Dichlorobenzoyl)benzoic acid 95%

Supplier: Ambeed
Warning

A227132-1G A227132-250MG
77165-424EA 178.19 USD
77165-424 77165-426
2-(3,4-Dichlorobenzoyl)benzoic acid 95%
2-(3,4-Dichlorobenzoyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₈Cl₂O₃
MW: 295.11 g/mol
MDL Number: MFCD00156968
CAS Number: 52187-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR