(3-Chloro-4-methoxyphenyl)methanamine hydrochloride 98%

Supplier: Ambeed
Danger

A227054-10G A227054-25G A227054-5G
77193-024EA 20 USD
77193-024 77193-026 77193-028
(3-Chloro-4-methoxyphenyl)methanamine hydrochloride 98%
(3-Chloro-4-methoxyphenyl)methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₁Cl₂NO
MDL Number: MFCD01211460
CAS Number: 41965-95-1
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR