2-Phenyl-1,2,3,4-tetrahydroisoquinoline 95%
Supplier: Ambeed
|
Warning
|
A226705-100MG
A226705-250MG
A226705-1G
77165-402EA
130
USD
77165-402
77165-404
77892-366
2-Phenyl-1,2,3,4-tetrahydroisoquinoline 95%
2-Phenyl-1,2,3,4-tetrahydroisoquinoline
Precaución: for Research Use Only
Formula:
C₁₅H₁₅N |
MDL Number:
MFCD18452288 CAS Number: 3340-78-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...