tert-Butyl ((3R,4S)-4-aminotetrahydrofuran-3-yl)carbamate 95%

Supplier: Ambeed
Warning

A226613-250MG A226613-100MG
77165-400EA 318.27 USD
77165-400 77683-657
tert-Butyl ((3R,4S)-4-aminotetrahydrofuran-3-yl)carbamate 95%
tert-Butyl ((3R,4S)-4-aminotetrahydrofuran-3-yl)carbamate

Precaución: For research use only.

Formula: C₉H₁₈N₂O₃
MW: 202.25 g/mol
MDL Number: MFCD27956984
CAS Number: 1628794-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR