Pyrrolidin-3-ylmethanol 95%
Supplier: Ambeed
|
Warning
|
A226516-1G
A226516-250MG
A226516-25G
A226516-5G
77165-396EA
20
USD
77165-396
77165-398
77892-362
77892-363
Pyrrolidin-3-ylmethanol 95%
Pyrrolidin-3-ylmethanol
Precaución: for Research Use Only
Formula:
C5H11NO |
MDL Number:
MFCD07773080 CAS Number: 5082-74-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...