2-Chloro-3-pyridineboronic acid pinacol ester 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-Chloropyridine-3-boronic acid pinacol ester
A226297-10G
A226297-1G
A226297-250MG
A226297-25G
A226297-5G
77165-384EA
52.53
USD
77165-384
77165-386
77165-388
77165-390
77165-392
2-Chloro-3-pyridineboronic acid pinacol ester 98%
2-Chloro-3-pyridineboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₁H₁₅BClNO₂ MW: 239.51 g/mol |
MDL Number:
MFCD04039873 CAS Number: 452972-11-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...