1,3,5-Trimethyl-2-phenoxybenzene 95%

Supplier: Ambeed
Danger

A226071-100MG A226071-1G A226071-250MG
77193-002EA 48 USD
77193-002 77193-004 77193-006
1,3,5-Trimethyl-2-phenoxybenzene 95%

Precaución: for Research Use Only

Formula: C₁₅H₁₆O
MDL Number: MFCD00623638
CAS Number: 61343-87-1
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR