7-(3-Chloropropoxy)-4-((2,4-dichloro-5-methoxyphenyl)amino)-6-methoxyquinoline-3-carbonitrile 97%
Supplier: Ambeed
|
Warning
|
A226035-100MG
A226035-1G
A226035-250MG
A226035-5G
77165-374EA
24
USD
77165-374
77165-376
77165-378
77165-380
7-(3-Chloropropoxy)-4-((2,4-dichloro-5-methoxyphenyl)amino)-6-methoxyquinoline-3-carbonitrile 97%
7-(3-Chloropropoxy)-4-((2,4-dichloro-5-methoxyphenyl)amino)-6-methoxyquinoline-3-carbonitrile
Precaución: for Research Use Only
Formula:
C₂₁H₁₈Cl₃N₃O₃ |
MDL Number:
MFCD09833975 CAS Number: 380844-49-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...