5-Bromo-8-hydroxyquinoline-7-carboxylic acid 95%

Supplier: Ambeed
Warning

A225944-100MG A225944-1G A225944-250MG
77165-368EA 160.68 USD
77165-368 77165-370 77165-372
5-Bromo-8-hydroxyquinoline-7-carboxylic acid 95%
5-Bromo-8-hydroxyquinoline-7-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₆BrNO₃
MW: 268.06 g/mol
MDL Number: MFCD18377724
CAS Number: 205040-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR