4-Bromo-5-methyl-1H-pyrazol-3(2H)-one 97%

Supplier: Ambeed
Warning

A225921-100MG A225921-10G A225921-250MG
77192-996EA 40.17 USD
77192-996 77192-998 77193-000
4-Bromo-5-methyl-1H-pyrazol-3(2H)-one 97%
4-Bromo-5-methyl-1H-pyrazol-3(2H)-one

Precaución: for Research Use Only

Formula: C₄H₅BrN₂O
MW: 177.00 g/mol
MDL Number: MFCD16620112
CAS Number: 89363-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR