Ethyl 4-cyanopiperidine-4-carboxylate hydrochloride 98%

Supplier: Ambeed
Warning

A225916-100MG A225916-1G A225916-250MG
77165-362EA 109.18 USD
77165-362 77165-364 77165-366
Ethyl 4-cyanopiperidine-4-carboxylate hydrochloride 98%
Ethyl 4-cyanopiperidine-4-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₅ClN₂O₂
MW: 218.68 g/mol
MDL Number: MFCD24387236
CAS Number: 123730-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR