NVP-BSK805 ≥98%

Supplier: Ambeed
Warning

Synonyms: 4-(2,6-difluoro-4-(3-(1-(piperidin-4-yl)-1H-pyrazol-4-yl)quinoxalin-5-yl)benzyl)morpholine, 8-[3,5-difluoro-4-(morpholin-4-ylmethyl)phenyl]-2-(1-piperidin-4-yl-1H-pyrazol-4-yl)quinoxaline, BSK 805

A225740-1MG A225740-2MG A225740-5MG
77192-990EA 21 USD
77192-990 77192-992 77192-994
NVP-BSK805 ≥98%
NVP-BSK805

Precaución: for Research Use Only

Formula: C₂₇H₂₈F₂N₆O
MDL Number: MFCD22419018
CAS Number: 1092499-93-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR