2-Oxa-5-azabicyclo[4.1.0]heptane hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A225246-1G
A225246-250MG
A225246-100MG
77165-346EA
742.63
USD
77165-346
77165-348
77683-655
2-Oxa-5-azabicyclo[4.1.0]heptane hydrochloride 95%
2-Oxa-5-azabicyclo[4.1.0]heptane hydrochloride
Precaución: For research use only.
Formula:
C₅H₁₀ClNO MW: 135.59 g/mol |
MDL Number:
MFCD20441759 CAS Number: 1354952-28-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...