(S)-1-Ethyl-3-(2-methoxy-4-(5-methyl-4-((1-(pyridin-3-yl)butyl)amino)pyrimidin-2-yl)phenyl)urea 98+%

Supplier: Ambeed
Warning

Synonyms: Lexibulin

A225027-10MG A225027-1MG A225027-5MG
77192-984EA 37.08 USD
77192-984 77192-986 77192-988
(S)-1-Ethyl-3-(2-methoxy-4-(5-methyl-4-((1-(pyridin-3-yl)butyl)amino)pyrimidin-2-yl)phenyl)urea 98+%
(S)-1-Ethyl-3-(2-methoxy-4-(5-methyl-4-((1-(pyridin-3-yl)butyl)amino)pyrimidin-2-yl)phenyl)urea

Precaución: for Research Use Only

Formula: C₂₄H₃₀N₆O₂
MW: 434.53 g/mol
MDL Number: MFCD18206787
CAS Number: 917111-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR