Methyl 4-amino-6-methoxypicolinate 97%

Supplier: Ambeed
Warning

A224513-100MG A224513-250MG
77192-980EA 207.03 USD
77192-980 77192-982
Methyl 4-amino-6-methoxypicolinate 97%
Methyl 4-amino-6-methoxypicolinate

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₃
MW: 182.17 g/mol
MDL Number: MFCD28137663
CAS Number: 1443759-42-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR