Benzyl (2-hydroxyethyl)(methyl)carbamate 98%

Supplier: Ambeed
Warning

A224424-1G A224424-25G A224424-5G
77165-310EA 11 USD
77165-310 77165-312 77165-314
Benzyl (2-hydroxyethyl)(methyl)carbamate 98%
Benzyl (2-hydroxyethyl)(methyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₃
MW: 209.24 g/mol
MDL Number: MFCD09909489
CAS Number: 67580-96-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR