Triallyl isocyanurate 97% stabilised
Supplier: Ambeed
|
Warning
|
Synonyms:
1,3,5-Triallyl-1,3,5-triazine-2,4,6(1H,3H,5H)-trione, Triallyl-s-triazine-2,4,6(1H,3H,5H)-trione, Isocyanuric Acid Triallyl Ester, 1,3,5-Triallyl-1,3,5-triazinane-2,4,6-trione
Stabilization:
Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
A224126-100G
A224126-10G
A224126-25G
A224126-500G
A224126-1000G
77192-964EA
8
USD
77192-964
77192-966
77192-968
77192-970
77550-456
Triallyl isocyanurate 97% stabilised
Triallyl isocyanurate
Precaución: for Research Use Only
Formula:
C₁₂H₁₅N₃O₃ |
MDL Number:
MFCD00006554 CAS Number: 1025-15-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...