(R)-tert-Butyl 3-(4-amino-3-(4-phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl)piperidine-1-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A224114-100MG
A224114-1G
A224114-250MG
A224114-5G
77192-956EA
14.42
USD
77192-956
77192-958
77192-960
77192-962
(R)-tert-Butyl 3-(4-amino-3-(4-phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl)piperidine-1-carboxylate 97%
(R)-tert-Butyl 3-(4-amino-3-(4-phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl)piperidine-1-carboxylate
Precaución: for Research Use Only
Formula:
C₂₇H₃₀N₆O₃ MW: 486.56 g/mol |
MDL Number:
MFCD28167899 CAS Number: 1022150-11-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...