4-((Dimethylamino)methyl)aniline 97%

Supplier: Ambeed
Warning

A223823-10G A223823-1G A223823-25G A223823-5G
77165-280EA 76 USD
77165-280 77165-282 77165-284 77165-286
4-((Dimethylamino)methyl)aniline 97%
4-((Dimethylamino)methyl)aniline

Precaución: for Research Use Only

Formula: C₉H₁₄N₂
MDL Number: MFCD00035953
CAS Number: 6406-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR