(S)-1-o-Tolylethanamine hydrochloride 95%

Supplier: Ambeed
Warning

A223667-100MG A223667-250MG A223667-1G
77165-276EA 15 USD
77165-276 77165-278 77892-356
(S)-1-o-Tolylethanamine hydrochloride 95%
(S)-1-o-Tolylethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₃N·ClH
MDL Number: MFCD11101268
CAS Number: 1332832-16-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR