1-Adamantanemethanol 98%

Supplier: Ambeed
Warning

Synonyms: 1-(Hydroxymethyl)adamantane, 1-(Hydroxymethyl)tricyclo[3.3.1.1(3,7)]decane, Adamantan-1-ylmethanol, 1-Adamantylmethanol

A223157-1G A223157-25G A223157-100G A223157-5G
77192-944EA 5.15 USD
77192-944 77192-946 77568-152 77568-154
1-Adamantanemethanol 98%
1-Adamantanemethanol

Precaución: For research use only.

Formula: C₁₁H₁₈O
MW: 166.26 g/mol
MDL Number: MFCD00074751
CAS Number: 770-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR