4-(Trifluoromethoxy)phenyl isothiocyanate 98%

Supplier: Ambeed
Danger

Synonyms: 4-(Trifluoromethoxy)phenylisothiocyanate

A223095-1G A223095-25G
77192-938EA 12.36 USD
77192-938 77192-940
4-(Trifluoromethoxy)phenyl isothiocyanate 98%
4-(Trifluoromethoxy)phenyl isothiocyanate

Precaución: for Research Use Only

Formula: C₈H₄F₃NOS
MW: 219.19 g/mol
MDL Number: MFCD00042412
CAS Number: 64285-95-6
UN: 1760
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR