2-((4-Chlorophenyl)ethynyl)benzaldehyde 97%

Supplier: Ambeed
Warning

A222189-100MG A222189-1G A222189-250MG A222189-5G
77192-910EA 13.39 USD
77192-910 77192-912 77192-914 77192-916
2-((4-Chlorophenyl)ethynyl)benzaldehyde 97%
2-((4-Chlorophenyl)ethynyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₉ClO
MW: 240.68 g/mol
MDL Number: MFCD28369390
CAS Number: 1251832-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR